Matrix™ DeepWell™ Storage Blocks
Actual product may vary
Thermo Scientific™

Matrix™ DeepWell™ Storage Blocks

Use these DeepWell™ storage blocks as mother plates when creating compound libraries from automated liquid handling instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityWell ShapeVolume (Metric) Well
4211-11Non-sterileRound1200 μL
4212-11SterileRound1200 μL
4221Non-sterileRound2200 μL
4222SterileRound2200 μL
4325Non-sterileSquare, Conical Bottom (V-well)240 μL
Catalog number 4211-11
Price (USD)
357.28
Case of 40
Sterility:
Non-sterile
Well Shape:
Round
Volume (Metric) Well:
1200 μL
Price (USD)
357.28
Case of 40
When creating compound libraries using automated liquid handling instruments, use Thermo Scientific™ Matrix™ DeepWell™ Storage Blocks as a mother plate. These strong blocks are constructed of polypropylene and are chemically resistant, meeting all standard microplate specifications. The unique chimney well design of the 1mL blcok reduces the risk of cross-contamination between wells during pipetting, while the 2mL block has cut-away sides for easy handling and placement on robotic systems.
Warranty: 90 days
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
For Use With (Equipment)ALPS Heat Sealers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
No. per Case40
No. per Pack40
Packaging40 Pk., 40 Cases
Product TypeDeepwell Storage Block
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
Volume (Metric) Working1000 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorNatural
FormatStandard
MaterialPolypropylene
No. of Wells96
Product LineNunc
SterilityNon-sterile
Surface TreatmentNon-treated
Volume (Metric) Well1200 μL
Well ShapeRound
Unit SizeCase of 40