Tricosane, 98%
Tricosane, 98%
Tricosane, 98%
Thermo Scientific Chemicals

Tricosane, 98%

CAS: 638-67-5 | C23H48 | 324.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 421600010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS638-67-5
IUPAC Nametricosane
Molecular FormulaC23H48
InChI KeyFIGVVZUWCLSUEI-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point47°C to 50°C
Appearance (Form)Crystals, flakes and/or chunks
GC>=97.5 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only