2,3,4-Trihydroxybenzaldehyde, 98%
2,3,4-Trihydroxybenzaldehyde, 98%
2,3,4-Trihydroxybenzaldehyde, 98%
Thermo Scientific Chemicals

2,3,4-Trihydroxybenzaldehyde, 98%

CAS: 2144-08-3 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 421790250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2144-08-3
IUPAC Name2,3,4-trihydroxybenzaldehyde
Molecular FormulaC7H6O4
InChI KeyCRPNQSVBEWWHIJ-UHFFFAOYSA-N
SMILESOC1=CC=C(C=O)C(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
GC>=97.5 %
Appearance (Color)Beige to light brown
Melting point159°C to 162°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only