2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
Thermo Scientific Chemicals

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%

CAS: 6846-50-0 | C16H30O4 | 286.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
Packaging:
Glass bottle
Catalog number 422000050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6846-50-0
IUPAC Name2,2,4-trimethyl-1-[(2-methylpropanoyl)oxy]pentan-3-yl 2-methylpropanoate
Molecular FormulaC16H30O4
InChI KeyOMVSWZDEEGIJJI-UHFFFAOYNA-N
SMILESCC(C)C(OC(=O)C(C)C)C(C)(C)COC(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only