Phenyltrimethylammonium hydroxide, 0.1 M in methanol
Phenyltrimethylammonium hydroxide, 0.1 M in methanol
Phenyltrimethylammonium hydroxide, 0.1 M in methanol
Thermo Scientific Chemicals

Phenyltrimethylammonium hydroxide, 0.1 M in methanol

CAS: 1899-02-1 | C9H15NO | 153.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 422021000
Price (USD)
180.65
Online exclusive
201.00
Save 20.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
180.65
Online exclusive
201.00
Save 20.35
Each
Chemical Identifiers
CAS1899-02-1
IUPAC NameN,N,N-trimethylanilinium hydroxide
Molecular FormulaC9H15NO
InChI KeyHADKRTWCOYPCPH-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless to light yellow
Strength0.09 to 0.11 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only