(R)-(-)-2-Amino-3-metil-1-butanol, 98 %, Thermo Scientific Chemicals
(R)-(-)-2-Amino-3-metil-1-butanol, 98 %, Thermo Scientific Chemicals
(R)-(-)-2-Amino-3-metil-1-butanol, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-2-Amino-3-metil-1-butanol, 98 %, Thermo Scientific Chemicals

CAS: 4276-09-9 | C5H13NO | 103.17 g/mol
Have Questions?
Número de catálogoCantidadEnvase
4224700505 gFrasco de vidrio
Número de catálogo 422470050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS4276-09-9
IUPAC Name(2R)-2-amino-3-methylbutan-1-ol
Molecular FormulaC5H13NO
InChI KeyNWYYWIJOWOLJNR-YFKPBYRVSA-N
SMILESCC(C)[C@@H](N)CO
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=97.5 %
Melting point32°C to 36°C
Specific optical rotation-15° to -18° (20°C, 589 nm) (c=10, EtOH)
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only