Xylene Cyanol, pure
Xylene Cyanol, pure
Xylene Cyanol, pure
Thermo Scientific Chemicals

Xylene Cyanol, pure

CAS: 4463-44-9 | C25H28N2NaO7S2 | 555.616 g/mol
数量:
25 g
50 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 422690250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4463-44-9
IUPAC Namesodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-6-hydroxybenzene-1,3-disulfonic acid hydride
Molecular FormulaC25H29N2NaO7S2
InChI KeyHLEQELPTFNOLFE-ZCTWALFCSA-N
SMILES[H-].[Na+].CCNC1=CC=C(C=C1C)C(=C1/C=CC(=NCC)C(C)=C1)\C1=CC(O)=C(C=C1S(O)(=O)=O)S(O)(=O)=O
さらに表示
Appearance (Color)Green to dark red to dark blue to purple
Appearance (Form)Powder
Infrared spectrumConforms
Dye content>=80 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only