Zinc phthalocyanine, 96%
Zinc phthalocyanine, 96%
Zinc phthalocyanine, 96%
Thermo Scientific Chemicals

Zinc phthalocyanine, 96%

CAS: 14320-04-8 | C32H16N8Zn | 577.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 422750250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS14320-04-8
IUPAC Namezinc(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide
Molecular FormulaC32H16N8Zn
InChI KeyPODBBOVVOGJETB-UHFFFAOYSA-N
SMILES[Zn++].[N-]1C2=NC3=C4C=CC=CC4=C([N-]3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<5 % (105°C)
UV spectrophotometry>=95 %
Appearance (Color)Blue to purple
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only