Acetone, 99.6%, ACS reagent
Acetone, 99.6%, ACS reagent
Acetone, 99.6%, ACS reagent
Thermo Scientific Chemicals

Acetone, 99.6%, ACS reagent

CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
4 L
25 L
Packaging:
View all
EcoSafPak™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 423240040
Price (USD)
-
Quantity:
4 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(in water) Passes test
Water=<0.5 % (K.F.)
Color scale=<10 APHA
GC>=99.5 %
Residue after evaporation=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only