Ammonium cerium(IV) nitrate, 99%, ACS reagent
Ammonium cerium(IV) nitrate, 99%, ACS reagent
Ammonium cerium(IV) nitrate, 99%, ACS reagent
Thermo Scientific Chemicals

Ammonium cerium(IV) nitrate, 99%, ACS reagent

CAS: 16774-21-3 | H8CeN8O18 | 548.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 423545000
Price (USD)
253.65
Online exclusive
282.00
Save 28.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
253.65
Online exclusive
282.00
Save 28.35
Each
Chemical Identifiers
CAS16774-21-3
IUPAC Nameλ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
Molecular FormulaCeH8N8O18
InChI KeyWIBGOERAEYJBOT-UHFFFAOYSA-N
SMILESN.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.01 %
Appearance (Form)Crystalline powder or crystals
Phosphate (PO4)=<0.02 %
Appearance (Color)Orange
Insoluble matter=<0.05 % (in dilute sulfuric acid)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only