Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Thermo Scientific Chemicals

Citric acid, ≥99.5%, ACS reagent, anhydrous

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
42356010010 kgPlastic drum
Catalog number 423560100
Price (USD)
-
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS77-92-9
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O7
InChI KeyKRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH>=99.5 %
Carboniz. sub. by hot H2SO4(tartrates, etc) Passes test
Chloride (Cl)=<10 ppm
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only