Cupferron
Cupferron
Cupferron
Thermo Scientific Chemicals

Cupferron

CAS: 135-20-6 | C6H9N3O2 | 155.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 423605000
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS135-20-6
IUPAC NameN-nitroso-N-phenylhydroxylamine amine
Molecular FormulaC6H9N3O2
InChI KeyNJVHCUNZAMFQNA-UHFFFAOYSA-N
SMILESN.ON(N=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Solubility(5 % in water) Passes test
Residue after ignition=<0.05 %
Additional infopackage usually contains small pouches
Appearance (Color)White to yellow-brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only