Iron(III) chloride hexahydrate, 97+%, ACS reagent
Iron(III) chloride hexahydrate, 97+%, ACS reagent
Iron(III) chloride hexahydrate, 97+%, ACS reagent
Thermo Scientific Chemicals

Iron(III) chloride hexahydrate, 97+%, ACS reagent

CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 423700025
Price (USD)
314.00
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
314.00
Each
Chemical Identifiers
CAS10025-77-1
IUPAC Nameiron(3+) hexahydrate trichloride
Molecular FormulaCl3FeH12O6
InChI KeyNQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to light brown
Appearance (Form)Adhering powder and/or lumps
Titration Iodometric97.0 to 102.0 %
Insoluble matter=<0.01 %
Phosphorous compounds (as PO4)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only