Iron(II) sulfate heptahydrate, 99+%, ACS reagent
Iron(II) sulfate heptahydrate, 99+%, ACS reagent
Iron(II) sulfate heptahydrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Iron(II) sulfate heptahydrate, 99+%, ACS reagent

CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
3 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 423730100
Price (USD)
1,054.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,241.00
Save 186.35 (15%)
Each
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
1,054.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,241.00
Save 186.35 (15%)
Each
Chemical Identifiers
CAS7782-63-0
IUPAC Nameλ²-iron(2+) heptahydrate sulfate
Molecular FormulaFeH14O11S
InChI KeySURQXAFEQWPFPV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Appearance (Color)Green to blue-green to light blue
Titration with KMnO4>=99.0 %
Insoluble matter=<0.01 %
Phosphate (PO4)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only