Magnesium sulfate heptahydrate, 98+%, ACS reagent
Magnesium sulfate heptahydrate, 98+%, ACS reagent
Magnesium sulfate heptahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, 98+%, ACS reagent

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic Drum
Catalog number 423900050
Price (USD)
-
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS10034-99-8
IUPAC Namemagnesium(2+) heptahydrate sulfate
Molecular FormulaH14MgO11S
InChI KeyWRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Titration Complexometric98.0 to 102.0 %
pH5.0 to 8.2 (5% soln. at 25°C)
Appearance (Color)White
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only