Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Thermo Scientific Chemicals

Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.

CAS: 67-56-1 | CH4O | 32.04 g/mol
数量:
500 mL
1 L
2.5 L
4 L
20 L
25 L
パッケージ:
すべて表示
EcoSafPak™
製品番号(カタログ番号) 423950040
価格(JPY)
-
数量:
4 L
パッケージ:
EcoSafPak™
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
さらに表示
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.8 %
Water=<0.1 % (K.F.)
Density(20/4°C) 0.791 to 0.793
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only