Methyl Red, ACS reagent
Methyl Red, ACS reagent
Methyl Red, ACS reagent
Thermo Scientific Chemicals

Methyl Red, ACS reagent

CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 423970100
Price (USD)
33.10
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.10
Each
Chemical Identifiers
CAS493-52-7
IUPAC Name2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
Molecular FormulaC15H15N3O2
InChI KeyCEQFOVLGLXCDCX-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to brown to purple
Clarity of alcohol solutionPasses test
Melting point179°C to 182°C
Visual transition intervalFrom pH 4.2 (pink) to pH 6.2 (yellow)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only