Molybdic acid, ACS reagent
Molybdic acid, ACS reagent
Molybdic acid, ACS reagent
Thermo Scientific Chemicals

Molybdic acid, ACS reagent

CAS: 7782-91-4 | H2MoO4 | 161.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 423981000
Price (USD)
78.65
Special offer
Online exclusive
Ends: 28-Dec-2026
92.70
Save 14.05
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
78.65
Special offer
Online exclusive
Ends: 28-Dec-2026
92.70
Save 14.05
Each
Chemical Identifiers
CAS7782-91-4
IUPAC Namedioxomolybdenumdiol
Molecular FormulaH2MoO4
InChI KeyVLAPMBHFAWRUQP-UHFFFAOYSA-L
SMILESO[Mo](O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.002 %
Phosphate (PO4)=<5 ppm
Sulfate (SO4)=<0.2 %
Heavy metals (as Pb)=<0.003 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only