Potassium hydroxide, ca. 85%, ACS reagent, pellets
Potassium hydroxide, ca. 85%, ACS reagent, pellets
Potassium hydroxide, ca. 85%, ACS reagent, pellets
Thermo Scientific Chemicals

Potassium hydroxide, ca. 85%, ACS reagent, pellets

CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
5 kg
25 kg
Catalog number 424140251
Price (USD)
-
Quantity:
25 kg
Request bulk or custom format
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl>=85 % (KOH)
Potassium carbonate=<2.0 % (K2CO3)
Sulfate (SO4)=<0.003 %
Heavy metals (ICP-OES)=<0.001 %
Iron (Fe)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only