Potassium phosphate, dibasic, 98+%, ACS reagent
Potassium phosphate, dibasic, 98+%, ACS reagent
Potassium phosphate, dibasic, 98+%, ACS reagent
Thermo Scientific Chemicals

Potassium phosphate, dibasic, 98+%, ACS reagent

CAS: 7758-11-4 | HK2O4P | 174.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic Drum
Catalog number 424190100
Price (USD)
1,110.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,307.00
Save 196.35 (15%)
Each
Quantity:
10 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
1,110.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,307.00
Save 196.35 (15%)
Each
Chemical Identifiers
CAS7758-11-4
IUPAC Namedipotassium hydrogen phosphate
Molecular FormulaHK2O4P
InChI KeyZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES[K+].[K+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Nitrogen compounds (as N)=<0.001 %
Sulfate (SO4)=<0.005 %
Heavy metals (ICP-OES)=<5 ppm
Titration excess HCl>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only