Sodium chloride, 99+%, ACS reagent
Sodium chloride, 99+%, ACS reagent
Sodium chloride, 99+%, ACS reagent
Thermo Scientific Chemicals

Sodium chloride, 99+%, ACS reagent

CAS: 7647-14-5 | ClNa | 58.44 g/mol
数量:
25 g
1 kg
5 kg
25 kg
パッケージ:
すべて表示
Amber Glass Bottle
製品番号(カタログ番号) 424290050
価格(JPY)
-
数量:
5 kg
パッケージ:
Amber Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
さらに表示
Magnesium (Mg)=<0.001 %
Potassium (K)=<0.005 %
Heavy metals (ICP-OES)=<5 ppm
pH5.0 to 9.0 (5 % solution at 25.0°C)
Insoluble matter=<0.005 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only