Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
Thermo Scientific Chemicals

Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 424370025
Price (USD)
410.65
Online exclusive
456.00
Save 45.35
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
410.65
Online exclusive
456.00
Save 45.35
Each
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<20 ppm
Sulfate (SO4)=<50 ppm
Appearance (Color)White
Appearance (Form)Powder or granular powder
Titration excess HCl>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH buffer
RUO – Research Use Only