Sodium tungstate dihydrate, 99+%, ACS reagent
Sodium tungstate dihydrate, 99+%, ACS reagent
Sodium tungstate dihydrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Sodium tungstate dihydrate, 99+%, ACS reagent

CAS: 10213-10-2 | Na2O42H2O | 329.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 424475000
Price (USD)
392.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
392.00
Each
Chemical Identifiers
CAS10213-10-2
IUPAC Namedisodium dioxotungstenbis(olate) dihydrate
Molecular FormulaH4Na2O6W
InChI KeyWPZFLQRLSGVIAA-UHFFFAOYSA-N
SMILESO.O.[Na+].[Na+].[O-][W]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.01 %
Appearance (Form)Crystalline powder or flakes
Chloride (Cl)=<50 ppm
Molybdenum (Mo)=<10 ppm
Sulfate (SO4)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only