Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent
Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent
Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 424571000
Price (USD)
54.30
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
54.30
Each
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Titration with HCl99.8 to 100.1 % (On dry basis)
Water=<2 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only