2-Methylsulphonylaniline hydrochloride, 95%
2-Methylsulphonylaniline hydrochloride, 95%
2-Methylsulphonylaniline hydrochloride, 95%
Thermo Scientific Chemicals

2-Methylsulphonylaniline hydrochloride, 95%

CAS: 205985-95-1 | C7H9NO2S | 171.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4248400252.5 gGlass bottle
Catalog number 424840025
Price (USD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS205985-95-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only