4-Methylsulphonylbenzylamine hydrochloride, 95%
4-Methylsulphonylbenzylamine hydrochloride, 95%
4-Methylsulphonylbenzylamine hydrochloride, 95%
Thermo Scientific Chemicals

4-Methylsulphonylbenzylamine hydrochloride, 95%

CAS: 4393-16-2 | C8H11NO2S | 185.241 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4249500252.5 gGlass bottle
Catalog number 424950025
Price (USD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98593-51-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Melting point269°C to 277°C
Titration with NaOH>=94 %
Appearance (Color)White to beige to light yellow-pink
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only