4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%
4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%
4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%
Thermo Scientific Chemicals

4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%

CAS: 17852-67-4 | C7H10N2O2S·HCl | 222.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Catalog number 424960025
Price (USD)
120.00
Each
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
120.00
Each
Chemical Identifiers
CAS17852-67-4
IUPAC Name(4-methanesulfonylphenyl)hydrazine
Molecular FormulaC7H10N2O2S
InChI KeyZHYAENSFCNMJQQ-UHFFFAOYSA-N
SMILESCS(=O)(=O)C1=CC=C(NN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only