Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Thermo Scientific™

Nalgene™ Polypropylene Analytical Funnels

Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopCapacity (Metric)Unit Size
4250-005554 mm41 mLCase of 36
4250-003535 mm10 mLCase of 36
4250-0035PK
also known as 4250-0035
35 mm10 mLPack of 12
4250-0055PK
also known as 4250-0055
54 mm41 mLPack of 12
4250-006563 mm64 mLCase of 36
4250-0065PK
also known as 4250-0065
63 mm64 mLPack of 12
4250-007576 mm114 mLCase of 36
4250-0075PK
also known as 4250-0075
76 mm114 mLPack of 6
4250-010098 mm254 mLCase of 24
4250-0100PK
also known as 4250-0100
98 mm254 mLPack of 4
4250-0160158 mm976 mLCase of 12
4250-0160PK
also known as 4250-0160
158 mm976 mLPack of 2
Catalog number 4250-0055
Price (USD)
177.17
Online exclusive
211.14
Save 33.97
Case of 36
Diameter (Metric)Top:
54 mm
Capacity (Metric):
41 mL
Unit Size:
Case of 36
Price (USD)
177.17
Online exclusive
211.14
Save 33.97
Case of 36

Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.

  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable

Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.

Specifications
AutoclavableAutoclavable
DescriptionFunnel Analytical PP 55mm
Diameter (Metric) Stem7 mm
Diameter (Metric)Top54 mm
Length (Metric) Stem61 mm
Quantity12/Pk.; 36/Cs.
Capacity (Metric)41 mL
MaterialPolypropylene
Unit SizeCase of 36