RadEye PRD4 and PRD-ER4 Personal Radiation Detector
El producto real puede variar
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
Thermo Scientific™
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
The Thermo Scientific™ RadEye™ PRD4 and PRD-ER4 Personal Radiation Detectors provide highly sensitive radiation detection capability with little to no nuisance alarms.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo de detectorDescripción
4250691711 detector de tasa de dosis baja, 1 detector de tasa de dosis altaPaquete de cargador/BTLE PRD-ER4
4250671261 detector de tasa de dosis bajaDetector personal de bolsillo PRD4
4250671271 detector de tasa de dosis baja, 1 detector de tasa de dosis altaDetector personal de bolsillo PRD-ER4
4250691601 detector de tasa de dosis bajaPaquete de cargador PRD4
4250691611 detector de tasa de dosis bajaPaquete de cargador/BTLE PRD4
4250691621 detector de tasa de dosis bajaKit PRD4
4250691631 detector de tasa de dosis bajaKit para vehículo PRD4
4250691701 detector de tasa de dosis baja, 1 detector de tasa de dosis altaPaquete de cargador PRD-ER4
4250691721 detector de tasa de dosis baja, 1 detector de tasa de dosis altaKit PRD-ER4
4250691731 detector de tasa de dosis baja, 1 detector de tasa de dosis altaKit para vehículo PRD-ER4
Número de catálogo 425069171
Precio (CLP)
-
Tipo de detector:
1 detector de tasa de dosis baja, 1 detector de tasa de dosis alta
Descripción:
Paquete de cargador/BTLE PRD-ER4
Achieve highly sensitive radiation detection with few nuisance alarms with Thermo Scientific™ RadEye™ PRD4 and PRD-ER4 Personal Radiation Detectors. RadEye PRD4 and PRD-ER4 radiation detectors save users valuable time adjudicating alarms while increasing scanning throughput. Using Natural Background Rejection (NBR) Technology, the RadEye PRD4 and PRD-ER4 radiation detectors ignore fluctuations in naturally occurring radiation (NORM) while analyzing the energy imbalance of artificial radiation. Additionally, these radiation detectors identify the discovered material in a manner configurable for your operation or user skills.
The RadEye PRD4 and PRD-ER4 Personal Radiation Detectors enable accurate dose rate measurements across the broader gamma spectrum, providing a greater assurance of personal safety. The neutron indication capability of the RadEye PRD4 provides an additional layer of detection critical to determining the level of radiation threat.
Efficiency in the field
  • Larger display
  • Neutron indication capability
  • 25% improved sensitivity over previous model
  • Linear high dose rate detector
Easy information, smarter decisions
  • No retraining or relearning for infrequent users
  • Get results automatically, without the need to press buttons
  • Quickly guides you through next steps after an alarm
  • Simple 4 button design
  • Comprehensive data neatly organized and presented on screen
Rugged and practical
  • Can be worn in holster or standard service belt
  • Small and lightweight
  • Long battery life
  • Drop resistant to 1.5m
  • IP65
  • Can be operated in extreme temperatures
View our online training here.
Especificaciones
DescripciónPaquete de cargador/BTLE PRD-ER4
Tipo de detector1 detector de tasa de dosis baja, 1 detector de tasa de dosis alta
IncluyeRadEye PRD-ER4, desktop charger, BTLE charging back, and batteries
Tipo de productoPersonal Radiation Detector
Unit SizeEach