2-(4-Pyridyl)malondialdehyde, 95%
2-(4-Pyridyl)malondialdehyde, 95%
2-(4-Pyridyl)malondialdehyde, 95%
Thermo Scientific Chemicals

2-(4-Pyridyl)malondialdehyde, 95%

CAS: 51076-46-1 | C8H7NO2 | 149.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4253400252.5 gGlass bottle
Catalog number 425340025
Price (USD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS51076-46-1
IUPAC Name2-(1,4-dihydropyridin-4-ylidene)propanedial
Molecular FormulaC8H7NO2
InChI KeyFZZFOXQLXQEYFV-UHFFFAOYSA-N
SMILESO=CC(C=O)=C1C=CNC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Assay>=94 %
Appearance (Color)Beige to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only