3,4-Epoxytetrahydrofuran, 96%
3,4-Epoxytetrahydrofuran, 96%
3,4-Epoxytetrahydrofuran, 96%
Thermo Scientific Chemicals

3,4-Epoxytetrahydrofuran, 96%

CAS: 285-69-8 | C4H6O2 | 86.09 g/mol
数量:
10 g
50 g
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 425530500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS285-69-8
IUPAC Name3,6-dioxabicyclo[3.1.0]hexane
Molecular FormulaC4H6O2
InChI KeyAIUTZIYTEUMXGG-UHFFFAOYNA-N
SMILESC1OCC2OC12
さらに表示
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=95 %
Refractive index1.4450 to 1.4490 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only