Ethyl glyoxalate, technical, ∼50% in toluene
Ethyl glyoxalate, technical, ∼50% in toluene
Ethyl glyoxalate, technical, ∼50% in toluene
Thermo Scientific Chemicals

Ethyl glyoxalate, technical, ∼50% in toluene

CAS: 924-44-7 | C4H6O3 | 102.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number 425760010
Price (USD)
275.65
Special offer
Online exclusive
Ends: 28-Dec-2026
324.00
Save 48.35
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
275.65
Special offer
Online exclusive
Ends: 28-Dec-2026
324.00
Save 48.35
Each
Chemical Identifiers
CAS924-44-7
IUPAC Nameethyl 2-oxoacetate
Molecular FormulaC4H6O3
InChI KeyDBPFRRFGLYGEJI-UHFFFAOYSA-N
SMILESCCOC(=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
Titration excess HCl>=48 % (Aldehyde function)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only