Sulfur trioxide N,N-dimethylformamide complex, 47+% active SO3
Sulfur trioxide N,N-dimethylformamide complex, 47+% active SO3
Sulfur trioxide N,N-dimethylformamide complex, 47+% active SO3
Thermo Scientific Chemicals

Sulfur trioxide N,N-dimethylformamide complex, 47+% active SO3

CAS: 29584-42-7 | C3H7NO4S | 153.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 425850250
Price (USD)
200.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
200.00
Each
Chemical Identifiers
CAS29584-42-7
IUPAC NameN,N-dimethylformamide; sulfonylideneoxidane
Molecular FormulaC3H7NO4S
InChI KeyAFDQGRURHDVABZ-UHFFFAOYSA-N
SMILESO=S(=O)=O.CN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to light brown
Infrared spectrumConforms
Titration with NaOH>=47.0 % (as SO3)
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only