Benzyl 4-oxo-1-piperidinecarboxylate, 97%, Thermo Scientific Chemicals
Benzyl 4-oxo-1-piperidinecarboxylate, 97%, Thermo Scientific Chemicals
Benzyl 4-oxo-1-piperidinecarboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl 4-oxo-1-piperidinecarboxylate, 97%, Thermo Scientific Chemicals

Catalog NumberQuantityPackaging
42596025025 mLGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 425960250
Price (JPY)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19099-93-5
IUPAC Namebenzyl 4-oxopiperidine-1-carboxylate
Molecular FormulaC13H15NO3
InChI KeyVZOVOHRDLOYBJX-UHFFFAOYSA-N
SMILESO=C(OCC1=CC=CC=C1)N1CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Powder or solid or liquid
Appearance (Color)Colorless to white to light yellow to light orange
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only