4-Chloro-2-fluoro-5-nitrotoluene, 98%
4-Chloro-2-fluoro-5-nitrotoluene, 98%
4-Chloro-2-fluoro-5-nitrotoluene, 98%
Thermo Scientific Chemicals

4-Chloro-2-fluoro-5-nitrotoluene, 98%

CAS: 18349-11-6 | C7H5ClFNO2 | 189.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4260100505 gGlass bottle
Catalog number 426010050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18349-11-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline mass
NMR (1H-NMR)Consistent with structure
Appearance (Color)Yellow to light brown
GC>=97.5 %
Melting point42°C to 45°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only