4,7-Phenanthroline, 98%
4,7-Phenanthroline, 98%
4,7-Phenanthroline, 98%
Thermo Scientific Chemicals

4,7-Phenanthroline, 98%

CAS: 230-07-9 | C12H8N2 | 180.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4261000101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 426100010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS230-07-9
IUPAC Name4,7-phenanthroline
Molecular FormulaC12H8N2
InChI KeyDATYUTWESAKQQM-UHFFFAOYSA-N
SMILESC1=CN=C2C=CC3=NC=CC=C3C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
NMR (1H-NMR)Consistent with structure
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point170°C to 174°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only