DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water
Thermo Scientific Chemicals

DL-1,4-Dithiothreitol, For Biochemistry, 1M solution in water

CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol
数量:
10 mL
50 mL
250 mL
製品番号(カタログ番号) 426380100
価格(JPY)
-
数量:
10 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
さらに表示
Appearance (Color)Clear colorless
Appearance (Form)Solution
UVat 500 nm A: =<0.05
Titration Iodimetric0.90 to 1.10 M
UV(neat)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only