(S)-(-)-3-(Dimethylamino)pyrrolidine, 97%, Thermo Scientific Chemicals
(S)-(-)-3-(Dimethylamino)pyrrolidine, 97%, Thermo Scientific Chemicals
(S)-(-)-3-(Dimethylamino)pyrrolidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(-)-3-(Dimethylamino)pyrrolidine, 97%, Thermo Scientific Chemicals

CAS: 132883-44-4 | C6H14N2 | 114.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4264500101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 426450010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS132883-44-4
IUPAC NameN,N-dimethylpyrrolidin-3-amine
Molecular FormulaC6H14N2
InChI KeyAVAWMINJNRAQFS-UHFFFAOYNA-N
SMILESCN(C)C1CCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Specific optical rotation-12° to -16° (20°C, 589 nm) (c=1% ethanol)
Refractive index1.4640 to 1.4660 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only