3-Guanidinopropionic acid, 97%
3-Guanidinopropionic acid, 97%
3-Guanidinopropionic acid, 97%
Thermo Scientific Chemicals

3-Guanidinopropionic acid, 97%

CAS: 353-09-3 | C4H9N3O2 | 131.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 426660010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS353-09-3
IUPAC Name3-[(diaminomethylidene)amino]propanoic acid
Molecular FormulaC4H9N3O2
InChI KeyKMXXSJLYVJEBHI-UHFFFAOYSA-N
SMILESNC(N)=NCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Titration with HClO4>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only