Isopropylmagnesium bromide, 3M solution in 2-MeTHF, AcroSeal™
Isopropylmagnesium bromide, 3M solution in 2-MeTHF, AcroSeal™
Isopropylmagnesium bromide, 3M solution in 2-MeTHF, AcroSeal™
Thermo Scientific Chemicals

Isopropylmagnesium bromide, 3M solution in 2-MeTHF, AcroSeal™

CAS: 920-39-8 | C3H7BrMg | 147.30 g/mol
製品番号(カタログ番号)数量パッケージ
426781000100 mLAcroSeal™ Glass Bottle
製品番号(カタログ番号) 426781000
価格(JPY)
-
数量:
100 mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS920-39-8
IUPAC Namebromo(propan-2-yl)magnesium
Molecular FormulaC3H7BrMg
InChI KeyUCJIVFREPMUZDC-UHFFFAOYSA-M
SMILESCC(C)[Mg]Br
さらに表示
Appearance (Form)Solution
Appearance (Color)Clear dark grey to dark brown
Titration Grignard2.85 to 3.20
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only