2-Bromo-3-thiophenecarboxylic acid, 96%
2-Bromo-3-thiophenecarboxylic acid, 96%
2-Bromo-3-thiophenecarboxylic acid, 96%
Thermo Scientific Chemicals

2-Bromo-3-thiophenecarboxylic acid, 96%

CAS: 24287-95-4 | C5H3BrO2S | 207.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4268400505 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 426840050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24287-95-4
IUPAC Name2-bromothiophene-3-carboxylic acid
Molecular FormulaC5H3BrO2S
InChI KeyRVSXMPCELBYUSF-UHFFFAOYSA-N
SMILESOC(=O)C1=C(Br)SC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=95 %
Infrared spectrumConforms
Appearance (Color)White to tan
Melting point178°C to 182°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only