Benzyloxyacetic Acid, 95%
Benzyloxyacetic Acid, 95%
Benzyloxyacetic Acid, 95%
Thermo Scientific Chemicals

Benzyloxyacetic Acid, 95%

CAS: 30379-55-6 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 427080010
Price (USD)
28.80
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
28.80
Each
Chemical Identifiers
CAS30379-55-6
IUPAC Name2-(benzyloxy)acetic acid
Molecular FormulaC9H10O3
InChI KeyGRZHHTYDZVRPIC-UHFFFAOYSA-N
SMILESOC(=O)COCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=94 %
Appearance (Color)Clear colorless to yellow
NMR (1H-NMR)Consistent with structure
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only