Di-tert-butylchlorophosphine, 96%, AcroSeal™
Di-tert-butylchlorophosphine, 96%, AcroSeal™
Di-tert-butylchlorophosphine, 96%, AcroSeal™
Thermo Scientific Chemicals

Di-tert-butylchlorophosphine, 96%, AcroSeal™

CAS: 13716-10-4 | C8H18ClP | 180.66 g/mol
Have Questions?
Catalog NumberQuantityPackaging
42728050050 mLAcroSeal™ Glass Bottle
Catalog number 427280500
Price (USD)
352.65
Special offer
Online exclusive
Ends: 28-Dec-2026
415.00
Save 62.35
Each
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
352.65
Special offer
Online exclusive
Ends: 28-Dec-2026
415.00
Save 62.35
Each
Chemical Identifiers
CAS13716-10-4
IUPAC Namedi-tert-butyl(chloro)phosphane
Molecular FormulaC8H18ClP
InChI KeyMCRSZLVSRGTMIH-UHFFFAOYSA-N
SMILESCC(C)(C)P(Cl)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=95 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only