Indole-2-carboxaldehyde, 97%
Indole-2-carboxaldehyde, 97%
Indole-2-carboxaldehyde, 97%
Thermo Scientific Chemicals

Indole-2-carboxaldehyde, 97%

CAS: 19005-93-7 | C9H7NO | 145.16 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 427510050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS19005-93-7
Appearance (Color)White to yellow to tan
Appearance (Form)Crystalline powder
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Melting point133°C to 143°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only