4-Hydroxy-3-methylbenzaldehyde, 98%
4-Hydroxy-3-methylbenzaldehyde, 98%
4-Hydroxy-3-methylbenzaldehyde, 98%
Thermo Scientific Chemicals

4-Hydroxy-3-methylbenzaldehyde, 98%

CAS: 15174-69-3 | C8H8O2 | 136.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4275500101 gGlass bottle
Catalog number 427550010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS15174-69-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to tan to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Melting point118°C to 120°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only