Methyl 4-imidazolecarboxylate, 98%
Methyl 4-imidazolecarboxylate, 98%
Methyl 4-imidazolecarboxylate, 98%
Thermo Scientific Chemicals

Methyl 4-imidazolecarboxylate, 98%

CAS: 17325-26-7 | C5H6N2O2 | 126.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 427630010
Price (USD)
124.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
124.00
Each
Chemical Identifiers
CAS17325-26-7
IUPAC Namemethyl 1H-imidazole-5-carboxylate
Molecular FormulaC5H6N2O2
InChI KeyDVLGIQNHKLWSRU-UHFFFAOYSA-N
SMILESCOC(=O)C1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point149°C to 158°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only