3-(N,N-Dimethylmyristylammonio)propanesulfonate, 97%
3-(N,N-Dimethylmyristylammonio)propanesulfonate, 97%
3-(N,N-Dimethylmyristylammonio)propanesulfonate, 97%
Thermo Scientific Chemicals

3-(N,N-Dimethylmyristylammonio)propanesulfonate, 97%

CAS: 14933-09-6 | C19H41NO3S | 363.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 427740250
Price (USD)
63.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.10
Save 11.45
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 28-Dec-2026
75.10
Save 11.45
Each
Chemical Identifiers
CAS14933-09-6
IUPAC Name3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
Molecular FormulaC19H41NO3S
InChI KeyBHATUINFZWUDIX-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Elemental Analysis60.57 to 64.96 % C
Infrared spectrumConforms
Elemental Analysis3.72 to 3.99 % N
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only