DL-3-Phenyllactic acid, 97%
DL-3-Phenyllactic acid, 97%
DL-3-Phenyllactic acid, 97%
Thermo Scientific Chemicals

DL-3-Phenyllactic acid, 97%

CAS: 828-01-3 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 427770010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS828-01-3
IUPAC Name2-hydroxy-3-phenylpropanoic acid
Molecular FormulaC9H10O3
InChI KeyVOXXWSYKYCBWHO-UHFFFAOYNA-N
SMILESOC(CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)White to almost white
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only