Phosphorus pentabromide, 95%
Phosphorus pentabromide, 95%
Phosphorus pentabromide, 95%
Thermo Scientific Chemicals

Phosphorus pentabromide, 95%

CAS: 7789-69-7 | Br5P | 430.49 g/mol
Have Questions?
Catalog NumberQuantityPackaging
42827025025 gGlass bottle
Catalog number 428270250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7789-69-7
IUPAC Namepentabromo-λ⁵-phosphane
Molecular FormulaBr5P
InChI KeyQRKVRHZNLKTPGF-UHFFFAOYSA-N
SMILESBrP(Br)(Br)(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Color)Yellow to orange to red
Titration Argentometric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only