Methyltitanium(IV) triisopropoxide, 1M solution in THF, AcroSeal™
Methyltitanium(IV) triisopropoxide, 1M solution in THF, AcroSeal™
Methyltitanium(IV) triisopropoxide, 1M solution in THF, AcroSeal™
Methyltitanium(IV) triisopropoxide, 1M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Methyltitanium(IV) triisopropoxide, 1M solution in THF, AcroSeal™

CAS: 18006-13-8 | C10H24O3Ti | 240.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
428371000100 mLAcroSeal™ Glass bottle
Catalog number 428371000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18006-13-8
IUPAC Nametris(propan-2-ol) titanium methanide
Molecular FormulaC10H27O3Ti
InChI KeyQBLPAZNBFFURKB-UHFFFAOYSA-N
SMILES[CH3-].[Ti].CC(C)O.CC(C)O.CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Strength0.9 to 1.1 M
Appearance (Color)Yellow to orange
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only